siRNA Details
| Gene Symbol: PDXDC2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003610.1 | 16, 17 | 1566 | |||
| GenBank mRNA | AK292860.1 | - | 16, 17 | 1436 | ||
| AL833251.1 | - | 16, 17 | 1566 | |||
| BC063633.1 | - | 15, 16 | 1438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDXDC2P| Gene Symbol: PDXDC2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003610.1 | 16, 17 | 1566 | |||
| GenBank mRNA | AK292860.1 | - | 16, 17 | 1436 | ||
| AL833251.1 | - | 16, 17 | 1566 | |||
| BC063633.1 | - | 15, 16 | 1438 | |||