siRNA Details
| Gene Symbol: LOC100129345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033943.1 | 4 | 1497 | |||
| GenBank mRNA | BX161483.1 | - | 4 | 1497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129345| Gene Symbol: LOC100129345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033943.1 | 4 | 1497 | |||
| GenBank mRNA | BX161483.1 | - | 4 | 1497 | ||