siRNA Details
Gene Symbol: LOC100129940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292023.1 | NP_001278952.1 | 3 | 2500 | ||
GenBank mRNA | AK128707.1 | - | 3 | 2374 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100129940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292023.1 | NP_001278952.1 | 3 | 2500 | ||
GenBank mRNA | AK128707.1 | - | 3 | 2374 |