siRNA Details
| Gene Symbol: LOC101927770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110051.1 | 4 | 744 | |||
| GenBank mRNA | AK024093.1 | - | 4 | 744 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000424252.2 | - | 4 | 744 | Chr. 20 - 53552770 - 53575308 [+] on Build GRCh38 | |

LOC101927770
Genomic Map