siRNA Details
| Gene Symbol: CYYR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135516.1 | 3 | 273 | |||
| GenBank mRNA | BX114294.1 | - | 3 | 273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYYR1-AS1| Gene Symbol: CYYR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135516.1 | 3 | 273 | |||
| GenBank mRNA | BX114294.1 | - | 3 | 273 | ||