siRNA Details
| Gene Symbol: LOC101928372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110695.1 | 5 | 746 | |||
| GenBank mRNA | BC035240.1 | - | 5 | 746 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000356006.3 | - | 746 | Chr. 1 - 155627547 - 155629395 [-] on Build GRCh38 | ||

LOC101928372
Genomic Map