siRNA Details
| Gene Symbol: C9orf41-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121183.1 | 5 | 641 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000455609.1 | - | 5 | 638 | Chr. 9 - 74952968 - 74996293 [+] on Build GRCh38 | |

C9orf41-AS1
Genomic Map