siRNA Details
| Gene Symbol: LINC01562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127900.1 | - | 2 | 834 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000366181.2 | - | 830 | Chr. 5 - 176376243 - 176376645 [+] on Build GRCh38 | ||

LINC01562
Genomic Map