siRNA Details
| Gene Symbol: LINC00449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133664.1 | 1 | 113 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000366259.2 | - | 1 | 102 | Chr. 7 - 72893318 - 72893594 [-] on Build GRCh38 | |

LINC00449
Genomic Map