siRNA Details
Gene Symbol: LOC101927447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134582.1 | 2 | 1702 | |||
GenBank mRNA | BC036874.1 | - | 2 | 1688 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000458463.1 | - | 2 | 1689 | Chr. 22 - 43275955 - 43283830 [+] on Build GRCh38 |
