siRNA Details
| Gene Symbol: LOC101927447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134582.1 | 2 | 1839 | |||
| GenBank mRNA | BC036874.1 | - | 2 | 1825 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000458463.1 | - | 2 | 1826 | Chr. 22 - 43275955 - 43283830 [+] on Build GRCh38 | |

LOC101927447
Genomic Map