siRNA Details
Gene Symbol: LINC01285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057519.1 | - | 7 | 670 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000412422.6 | - | 7 | 670 | Chr. X - 118839556 - 118857886 [+] on Build GRCh38 |
