siRNA Details
| Gene Symbol: LOC441178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117092.1 | 4 | 582 | |||
| GenBank mRNA | AK056600.1 | - | 4 | 582 | ||
| AL832737.1 | - | 6 | 1913 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000400831.2 | - | 4 | 582 | Chr. 6 - 167679626 - 167683787 [-] on Build GRCh38 | |

LOC441178
Genomic Map