siRNA Details
Gene Symbol: LOC441178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117092.1 | 4 | 2221 | |||
GenBank mRNA | AK056600.1 | - | 4 | 2221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000400831.2 | - | 4 | 2221 | Chr. 6 - 167679626 - 167683787 [-] on Build GRCh38 |
