siRNA Details
| Gene Symbol: LINC00272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034131.1 | 3 | 507 | |||
| GenBank mRNA | AF508909.1 | - | 3 | 507 | ||
| BC140780.1 | - | 3 | 357 | 9021648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00272| Gene Symbol: LINC00272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034131.1 | 3 | 507 | |||
| GenBank mRNA | AF508909.1 | - | 3 | 507 | ||
| BC140780.1 | - | 3 | 357 | 9021648 | ||