siRNA Details
Gene Symbol: LOC101928673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125960.1 | 2 | 1082 | |||
GenBank mRNA | AK130460.1 | - | 2 | 1083 | ||
BC136808.1 | - | 2 | 1075 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125960.1 | 2 | 1082 | |||
GenBank mRNA | AK130460.1 | - | 2 | 1083 | ||
BC136808.1 | - | 2 | 1075 |