siRNA Details
| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033376.1 | 4 | 580 | |||
| GenBank mRNA | AY927444.1 | - | LIT3473 AY927444 |
4 | 578 | |
| AY927447.1 | - | LIT3475 AY927447 |
4 | 543 | ||
| AY927448.1 | - | LIT3476 AY927448 |
4 | 422 | ||

LINC00222
Genomic Map