siRNA Details
| Gene Symbol: LINC00222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033376.1 | 2 | 313 | |||
| GenBank mRNA | AY927443.1 | - | LIT3472 AY927443 |
2 | 314 | |
| AY927444.1 | - | LIT3473 AY927444 |
2 | 311 | ||
| AY927445.1 | - | 2 | 281 | |||
| AY927446.1 | - | LIT3474 AY927446 |
2 | 314 | ||
| AY927447.1 | - | LIT3475 AY927447 |
2 | 276 | ||
| AY927448.1 | - | LIT3476 AY927448 |
2 | 155 | ||
| AY927449.1 | - | LIT3477 AY927449 |
2 | 149 | ||

LINC00222
Genomic Map