siRNA Details
| Gene Symbol: BVES-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037157.1 | 4 | 836 | |||
| GenBank mRNA | AL137368.1 | - | 4 | 838 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000369122.7 | - | 4 | 836 | Chr. 6 - 105137687 - 105169945 [+] on Build GRCh38 | |

BVES-AS1
Genomic Map