siRNA Details
| Gene Symbol: LINC00632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028345.2 | 2 | 645 | |||
| GenBank mRNA | DA529197.1 | - | 1 | 268 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000607004.5 | - | 2 | 637 | Chr. X - 140709767 - 140712229 [+] on Build GRCh38 | |

LINC00632
Genomic Map