siRNA Details
| Gene Symbol: LOC100133077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121583.1 | 1 | 116 | |||
| GenBank mRNA | AK128414.1 | - | 1 | 116 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000371390.1 | - | 1 | 116 | Chr. 9 - 137867925 - 137892570 [-] on Build GRCh38 | |

LOC100133077
Genomic Map