siRNA Details
| Gene Symbol: LINC01270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034124.1 | 5 | 1941 | |||
| GenBank mRNA | AK090605.1 | - | 5 | 1941 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01270| Gene Symbol: LINC01270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034124.1 | 5 | 1941 | |||
| GenBank mRNA | AK090605.1 | - | 5 | 1941 | ||