siRNA Details
| Gene Symbol: LINC01270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034124.1 | 5 | 1238 | |||
| GenBank mRNA | AK090605.1 | - | 5 | 1238 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000371639.7 | - | 1238 | Chr. 20 - 50292749 - 50298391 [+] on Build GRCh38 | ||

LINC01270
Genomic Map