siRNA Details
| Gene Symbol: LINC00951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038887.1 | 2 | 2662 | |||
| GenBank mRNA | AK123643.1 | - | 2 | 2662 | ||
| BC132803.1 | - | 2 | 402 | |||
| BC132805.1 | - | 2 | 402 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000373171.2 | - | 1 | 93 | Chr. 6 - 40344345 - 40356006 [-] on Build GRCh38 | |

LINC00951
Genomic Map