siRNA Details
Gene Symbol: LOC100129029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135242.1 | 4 | 1152 | |||
GenBank mRNA | AK126351.1 | - | 4 | 1154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000342892.4 | - | 4 | 1152 | Chr. 2 - 158658337 - 158735002 [-] on Build GRCh38 |
