siRNA Details
Gene Symbol: LOC100129029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135242.1 | 2 | 341 | |||
GenBank mRNA | AK126351.1 | - | 2 | 343 | ||
BC039475.1 | - | 2 | 157 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100129029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135242.1 | 2 | 341 | |||
GenBank mRNA | AK126351.1 | - | 2 | 343 | ||
BC039475.1 | - | 2 | 157 |