siRNA Details
| Gene Symbol: LOC151121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122040.1 | 5 | 2181 | |||
| NR_122041.1 | 4 | 2093 | ||||
| NR_122042.1 | 4 | 2104 | ||||
| GenBank mRNA | AK056598.1 | - | 5 | 2138 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000375987.3 | - | 2138 | Chr. 2 - 129242388 - 129242693 [-] on Build GRCh38 | ||

LOC151121
Genomic Map