siRNA Details
| Gene Symbol: LOC400997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122074.1 | 4 | 2906 | |||
| GenBank mRNA | AK125994.1 | - | 3 | 2724 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000376593.2 | - | 2 | 2724 | Chr. 2 - 111098352 - 111102078 [-] on Build GRCh38 | |

LOC400997
Genomic Map