siRNA Details
Gene Symbol: LINC01123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046110.1 | 2, 3 | 1454 | |||
GenBank mRNA | AK056084.1 | - | 2, 3 | 1371 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419296.1 | - | 2, 3 | 1371 | Chr. 2 - 109987063 - 109995281 [+] on Build GRCh38 |
