siRNA Details
| Gene Symbol: LINC01257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026670.2 | 4 | 1430 | |||
| GenBank mRNA | AL137487.1 | - | 4 | 1430 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000376678.6 | - | 4 | 1430 | Chr. 12 - 131165011 - 131212930 [+] on Build GRCh38 | |

LINC01257
Genomic Map