siRNA Details
Gene Symbol: FLJ26850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027257.1 | 2 | 588 | |||
GenBank mRNA | AK130360.1 | - | 2 | 679 | ||
BC130542.1 | - | 2 | 588 | |||
BC130544.1 | - | 2 | 588 | |||
HY015479.1 | - | 43 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000527209.1 | - | 2 | 679 | Chr. 19 - 50050589 - 50066793 [+] on Build GRCh38 |
