siRNA Details
| Gene Symbol: LOC284788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027089.2 | 1 | 433 | |||
| NR_027090.1 | 1 | 433 | ||||
| GenBank mRNA | BC033532.1 | - | 1 | 433 | ||
| BC144090.1 | - | 1 | 305 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000377121.1 | - | 1 | 433 | Chr. 20 - 22400351 - 22420643 [-] on Build GRCh38 | |

LOC284788
Genomic Map