siRNA Details
| Gene Symbol: LINC01556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103538.1 | 1 | 461 | |||
| GenBank mRNA | BX118328.1 | - | 1 | 461 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000377186.3 | - | 2 | 173 | Chr. 6 - 28943877 - 28944537 [+] on Build GRCh38 | |

LINC01556
Genomic Map