siRNA Details
| Gene Symbol: LINC01291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125960.1 | - | 1 | 1851 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000377469.1 | - | 1 | 1851 | Chr. 2 - 74928239 - 74933024 [+] on Build GRCh38 | |

LINC01291
Genomic Map