siRNA Details
Gene Symbol: LINC01291 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125960.1 | - | 1 | 3044 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000377469.1 | - | 1 | 3044 | Chr. 2 - 74928239 - 74933024 [+] on Build GRCh38 |
