siRNA Details
| Gene Symbol: LINC01545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046101.1 | 3 | 233 | |||
| GenBank mRNA | BC038573.2 | - | 3 | 233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01545| Gene Symbol: LINC01545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046101.1 | 3 | 233 | |||
| GenBank mRNA | BC038573.2 | - | 3 | 233 | ||