siRNA Details
Gene Symbol: C7orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024607.2 | NP_001019778.1 | 2 | 289 | ||
GenBank mRNA | BG211856.1 | - | 2 | 290 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000624695.1 | - | 289 | Chr. - - [-] on Build |
