siRNA Details
Gene Symbol: LINC00654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015406.2 | 2 | 3765 | |||
GenBank mRNA | AL137476.1 | - | 1 | 2905 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000379053.4 | - | 3782 | Chr. 20 - 5500331 - 5504613 [-] on Build GRCh38 |
