siRNA Details
| Gene Symbol: LINC01460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135200.1 | 2 | 2289 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000379677.2 | - | 2289 | Chr. 2 - 27706763 - 27715732 [-] on Build GRCh38 | ||

LINC01460
Genomic Map