siRNA Details
| Gene Symbol: LOC400867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120405.1 | 3 | 731 | |||
| GenBank mRNA | AK127082.1 | - | 3 | 731 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000380931.6 | - | 731 | Chr. 21 - 38878506 - 38878916 [-] on Build GRCh38 | ||

LOC400867
Genomic Map