siRNA Details
Gene Symbol: LINC01446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038371.1 | 3 | 1061 | |||
GenBank mRNA | AK127870.1 | - | 3 | 1061 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000380970.2 | - | 1061 | Chr. 7 - 53779942 - 53811923 [-] on Build GRCh38 |
