siRNA Details
| Gene Symbol: LINC01446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038371.1 | 6 | 2235 | |||
| GenBank mRNA | AK127870.1 | - | 6 | 2235 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000380970.2 | - | 2235 | Chr. 7 - 53779942 - 53811923 [-] on Build GRCh38 | ||

LINC01446
Genomic Map