siRNA Details
| Gene Symbol: PP13004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF447883.1 | - | 1 | 120 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000381493.2 | - | 1 | 120 | Chr. 7 - 36079084 - 36085730 [+] on Build GRCh38 | |

PP13004
Genomic Map