siRNA Details
| Gene Symbol: LINC01531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040046.1 | 2 | 2124 | |||
| GenBank mRNA | AK128099.1 | - | 2 | 2124 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000536898.5 | - | 2 | 2124 | Chr. 19 - 35405607 - 35416840 [+] on Build GRCh38 | |

LINC01531
Genomic Map