siRNA Details
| Gene Symbol: FAM99A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026643.1 | 3 | 1176 | |||
| GenBank mRNA | AK124823.1 | - | 1 | 1780 | ||
| CR627417.1 | - | 3 | 1203 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000382167.2 | - | 3 | 1178 | Chr. 11 - 1665595 - 1667855 [+] on Build GRCh38 | |

FAM99A
Genomic Map