siRNA Details
| Gene Symbol: C21orf62-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024622.1 | 3, 4 | 438 | |||
| NR_024623.1 | 3, 4 | 426 | ||||
| GenBank mRNA | AK096601.1 | - | 3, 4 | 426 | ||
| BC117397.1 | - | 3, 4 | 320 | |||
| BC117399.1 | - | 3, 4 | 332 | |||
| DA776905.1 | - | 3, 4 | 426 | |||

C21orf62-AS1
Genomic Map