siRNA Details
| Gene Symbol: LINC01296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122112.1 | 8 | 863 | |||
| GenBank mRNA | BX248778.1 | - | 8 | 863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01296| Gene Symbol: LINC01296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122112.1 | 8 | 863 | |||
| GenBank mRNA | BX248778.1 | - | 8 | 863 | ||