siRNA Details
| Gene Symbol: KRBOX1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122033.1 | 2 | 1119 | |||
| GenBank mRNA | AK057338.1 | - | 2 | 1119 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000447834.1 | - | 2 | 1119 | Chr. 3 - 42934252 - 42936785 [-] on Build GRCh38 | |

KRBOX1-AS1
Genomic Map