siRNA Details
| Gene Symbol: ZNF337-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126465.1 | 4 | 498 | |||
| GenBank mRNA | BC034569.1 | - | 5 | 3660 | ||
| BM907163.1 | - | 4 | 490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF337-AS1| Gene Symbol: ZNF337-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126465.1 | 4 | 498 | |||
| GenBank mRNA | BC034569.1 | - | 5 | 3660 | ||
| BM907163.1 | - | 4 | 490 | |||