siRNA Details
Gene Symbol: IPO5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103741.1 | 3, 4 | 375 | |||
NR_103742.1 | 2, 3 | 300 | ||||
GenBank mRNA | AK023040.1 | - | 3, 4 | 354 | ||
AK056739.1 | - | 1, 2 | 1014 | |||
DA183233.1 | - | 2, 3 | 265 | |||
DA185451.1 | - | 3, 4 | 360 | |||
DA510220.1 | - | 3, 4 | 382 | |||
DA723810.1 | - | 2, 3 | 300 |
